Learn More
colony stimulating factor 3 (granulocyte), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CSF3 protein.
Supplier: Abnova Corporation H00001440D01P
Description
The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes. The active protein is found extracellularly. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq
Sequence: MAGPATQSPMKLMALQLLLWHSALWTVQEATPLGPASSLPQSFLLKCLEQVRKIQGDGAALQEKLVSECATYKLCHPEELVLLGHSLGIPWAPLSSCPSQALQLAGCLSQLHSGLFLYQGLLQALEGISPELGPTLDTLQLDVADFATTIWQQMEELGMAPALQPTQGAMPAFASAFQRRAGGVLVASHLQSFLEVSYRVLRHLAQPSpecifications
colony stimulating factor 3 (granulocyte) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human CSF3 protein. | |
CSF3 | |
G-CSF/GCSF/MGC45931 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000759 | |
CSF3 | |
CSF3 (NP_000750.1, 1 a.a. ∼ 207 a.a) full-length human protein. | |
100 μg | |
Cell Cycle | |
1440 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.