Learn More
Csk Mouse, Unlabeled, Clone: 52, BD
Mouse Monoclonal Antibody
Specifications
Antigen | Csk |
---|---|
Clone | 52 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The csk gene encodes a protein kinase that phosphorylates several Src family kinases including p60(c-src), p59(fyn), p53/56(lyn), p56(lck), and p62(yes). This phosphorylation occurs specifically at the carboxy-terminal tyrosine, Y527. Csk (c-src kinase) was identified based on its ability to phosphorylate exogenous Src in mammalian src-deficient cells. Establishment of a mouse csk- cell line revealed that phosphorylation of Y527 in Src was reduced 20-50%. This correlated with an increase of Src kinase activity, thus indicating Csk is a negative regulator of src. Csk-deficient cells also show an elevation in the kinase activity of p59(fyn) and p53/56(lyn). These data suggest that Csk may be a universal negative regulator of Src family kinases in vivo.
Host Species: Mouse
Clone: 52
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Rat Csk aa. 1-156
Formula Weight [Chemical]: 50kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Csk | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
52 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Rat Csk aa. 1-156 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.