Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
casein kinase 1, gamma 2, Mouse, Clone: 2E6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CSNK1G2.
Supplier: Abnova Corporation H00001455M02
Description
Sequence: MDFDKKGGKGETEEGRRMSKAGGGRSSHGIRSSGTSSGVLMVGPNFRVGKKIGCGNFGELRLGKNLYTNEYVAIKLEPIKSRAPQLHLEYRFYKQLSATEGVPQVYYFGPCGNYNAMVLELLGPSLEDLFDLCDRTFTLKTVLMIAIQLITRMEYVHTKSLIYRDVKPENFLVGRPGTKRQHAIHIIDFGLAKEYIDPETKKHIPYREHKSLTGTARYMSINTHLGKEQSRRDDLEALGHMFMYFLRGSLPWQGLKADTLKERYQKIGDTKRATPIEVLCENFPEEMATYLRYVRRLDFFEKPDYDYLRKLFTDLFDRSGFVFDYEYDWAGKPLPTPIGTVHTDLPSQPQLRDKTQPHSKNQALNSTNGELNADDPTAGHSNAPITAPAEVEVADETKCCCFFKRRKRKSLQRHKSpecifications
casein kinase 1, gamma 2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC020972 | |
CSNK1G2 | |
CSNK1G2 (AAH20972.1, 1 a.a. ∼ 415 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Kinases | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
2E6 | |
Mouse monoclonal antibody raised against a full length recombinant CSNK1G2. | |
CSNK1G2 | |
CK1g2 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
1455 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction