Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

structural maintenance of chromosomes 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-713 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-713 Supplier Abnova Corporation Supplier No. H00009126B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CSPG6 protein.

This gene belongs to the SMC3 subfamily of SMC proteins. The encoded protein occurs in certain cell types as either an intracellular, nuclear protein or a secreted protein. The nuclear form, known as structural maintenance of chromosomes 3, is a component of the multimeric cohesin complex that holds together sister chromatids during mitosis, enabling proper chromosome segregation. Post-translational modification of the encoded protein by the addition of chondroitin sulfate chains gives rise to the secreted proteoglycan bamacan, an abundant basement membrane protein. [provided by RefSeq

Sequence: MYIKQVIIQGFRSYRDQTIVDPFSSKHNVIVGRNGSGKSNLFYAIQFVLSDEFSHLRPEQRLALLHEGTGPRVISAFVEIIFDNSDNRLPIDKEEVSLRRVIGAKKDQYFLDKKMVTKNDVMNLLESAGFSRSNPYYIVKQGKINQMATAPDSQRLKLLREVAGTRVYDERKEESISLMKETEGKREKINELLKYIEERLHTLEEEKEELAQYQKWDKMRRALEYTIYNQELNETRAKLDELSAKRETSGEKSRQLRDAQQDARDKMEDIERQVRELKTKISAMKEEKEQLSAERQEQIKQRTKLELKAKDLQDELAGNSEQRKRLLKERQKLLEKIEEKQKELAETEPKFNSVKEKEERGIARLAQATQERTDLYAKQGRGSQFTSKEERDKWIKKELKSLDQAINDKKRQIAAIHKDLEDTEANKEKNLEQYNKLDQDLNEVKARVEELDRKYYEVKNKKDELQSERNYLWREENAEQQALAAKREDLEKKQQLLRAATGKAILNGIDSINKVLDHFRRKGINQHVQNGYHGIVMNNFECEPAFYTCVEVTAGNRLFYHIVDSDEVSTKILMEFNKMNLPGEVTFLPLNKLDVRDTAYPETNDAIPMISKLRYNPRFDKAFKHVFGKTLICRSMEVSTQLARAFTMDCITLEGDQVSHRGALTGGYYDTRKSRLELQKDVRKAEEELGELEAKLNENLRRNIERINNEIDQLMNQMQQIETQQRKFKASRDSILSEMKMLKEKRQQSEKTFMPKQRSLQSLEASLHAMESTRESLKAELGTDLLSQLSLEDQKRVDALNDEIRQLQQENRQLLNERIKLEGIITRVETYLNENLRKRLDQVEQELNELRETEGGTVLTATTSELEAINKRVKDTMARSEDLDNSIDKTEAGIKELQKSMERWKNMEKEHMDAINHDTKELEKMTNRQGMLLKKKEECMKKIRELGSLPQEAFEKYQTLSLKQLFRKLEQCNTELKKYSHVNKKALDQFVNFSEQKEKLIKRQEELDRGYKSIMELMNVLELRKYEAIQLTFKQVSKNFSEVFQKLVPGGKATLVMKKGDVEGSQSQDEGEGSGESERGSGSQSSVPSVDQFTGVGIRVSFTGKQGEMREMQQLSGGQKSLVALALIFAIQKCDPAPFYLFDEIDQALDAQHRKAVSDMIMELAVHAQFITTTFRPELLESADKFYGVKFRNKVSHIDVITAEMAKDFVEDDTTHG

Specifications

Antigen structural maintenance of chromosomes 3
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CSPG6 protein.
Formulation No additive
Gene SMC3
Gene Accession No. BC047324
Gene Alias BAM/BMH/CDLS3/CSPG6/HCAP/SMC3L1
Gene Symbols SMC3
Host Species Mouse
Immunogen CSPG6 (AAH47324.1, 1 a.a. ∼ 1217 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9126
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.