Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAIP12 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535134
Description
CSRNP2 binds to the consensus sequence 5′-AGAGTG-3′ and has transcriptional activator activity. May play a role in apoptosis.
Specifications
TAIP12 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9H175 | |
CSRNP2 | |
KLH conjugated synthetic peptide between 509-538 amino acids from the C-terminal region of human CSRNP2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
CSRNP2 | |
CSRNP2; C12orf22; FAM130A1; TAIP12; Cysteine/serine-rich nuclear protein 2; Protein FAM130A1; TGF-beta-induced apoptosis protein 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81566 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction