Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cancer/testis antigen 2, Mouse, Clone: 3H1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CTAG2.
Supplier: Abnova Corporation H00030848M04
Description
Sequence: RDAAPLPRPGAVLKDFTVSGNLLFMSVRDQDREGAGRMRVVGWGLGSASPEGQKARDLRTPKHKVSEQRPGTPGPPPPEGAQGDGCRGVAFNVMFSAPHISpecifications
cancer/testis antigen 2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_020994 | |
CTAG2 | |
CTAG2 (NP_066274, 111 a.a. ∼ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
30848 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
ELISA, Western Blot | |
3H1 | |
Mouse monoclonal antibody raised against a partial recombinant CTAG2. | |
CTAG2 | |
CAMEL/ESO2/LAGE-1/LAGE-2b/MGC138724/MGC3803 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction