Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTCF Mouse, Unlabeled, Clone: 48, BD

Catalog No. BDB612148 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612148 50 μg
BDB612149 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612148 Supplier BD Biosciences Supplier No. 612148
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The family of nucleic acid-binding C2H2 type zinc finger transcription factors is divided into two classes. One class consists of small proteins (Gli1, Krox-20, WT1, Egr-1, and Sp1) with conserved zinc finger clusters of 3 to 5 units, while the other class (ZNF91, ZNF74, ZFP37, CTCF) can contain more than 10 zinc finger clusters. CTCF is a ubiquitously expressed, highly conserved transcription factor that contains 10 C2H2- and 1 C2H-type zinc-finger motifs. CTCF binds to and represses transcription at the promoter-proximal regions of c-myc oncogenes,
while CTCF can bind to and activate transcription at the promoter for amyloid β. Another mechanism of CTCF-mediated repression may include binding to insulator regions between enhancers and promoters resulting in enhancer blocking. In addition, CTCF gene may be a candidate tumor suppressor gene, since it localizes to a narrow cancer-associated chromosome region, and has been shown to have tumor-specific rearrangements in breast cancer patients. Thus, CTCF transcriptional regulation may be important for cell cycle progression, differentiation, apoptosis, and tumorigenesis.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CTCF
Applications Western Blot
Classification Monoclonal
Clone 48
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CTCF aa. 184-290
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.