Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTCF Rabbit anti-Human, Rat, Canine, Mouse, Primate, Polyclonal, MilliporeSigma™

Catalog No. 07729MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-729-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-729-MI Supplier MilliporeSigma Supplier No. 07729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-CTCF Antibody (Rabbit Polyclonal Antibody) validated in Chimmunoprecipitation, western blot, ChIP-seq to detect CTCF also known as 11-zinc finger protein, CCCTC-binding factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTCF
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum containing 0.05% sodium azide and 30% glycerol.
Gene Symbols CTCF
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 659-675 (C-TNQPKQNQPTAIIQVED) of human CCCTC-binding factor (CTCF) with a N-terminal cysteine added for conjugation purposes.
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006565.2
Target Species Canine, Human, Mouse, Primate, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.