Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTDSPL2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524421
Description
This antibody is predicted to react with rat based on sequence homology.
Probable phosphatase.
Specifications
CTDSPL2 | |
Polyclonal | |
CTDSPL2 | |
HSPC129, CTD small phosphatase-like protein 2, CTDSP-like 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
329506, 51496 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q05D32, Q8BG15 | |
CTDSPL2 | |
KLH conjugated synthetic peptide between 52-80 amino acids from the N-terminal region of human CTDSPL2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction