Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CTGF/CCN2 C-Terminus Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB660500 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB660500 500 μg
MAB660SP 25 μg
MAB660100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB660500 Supplier R&D Systems Supplier No. MAB660500

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

CTGF/CCN2 Monoclonal specifically detects CTGF/CCN2 in Human∣Mouse samples. It is validated for Bioassay∣ELISA Development∣Immunohistochemistry∣Western Blot.

Specifications

Antigen CTGF/CCN2
Applications Bioassay, ELISA Development, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 88430
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001892.1
Gene Alias CCN2, CCN2IGFBP-8, connective tissue growth factor, CTGRP, Fisp12, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP-8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
Gene Symbols CTGF
Host Species Mouse
Immunogen E. coli-derived recombinant human CTGF Glu247-Ala349 Accession # NP_001892.1
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1490
Test Specificity Detects human CTGF C-teriminal peptide in direct ELISAs. Does not cross-react with recombinant human (rh) LDGF, rhNOV, rhPDGF, rhPlGF, rhVEGF 206 rhVEGF-B 186 rhVEGF-C, rhVEGF-D, or rhWISP-1.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.