Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
collagen triple helix repeat containing 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CTHRC1.
Supplier: Abnova Corporation H00115908A01
Description
CTHRC1 is specifically expressed in vascular calcifications of carotid artery lesions and may contribute to vascular remodeling of injured arteries (Pyagay et al., 2005 [PubMed 15618538]).[supplied by OMIM
Sequence: EIPKGKQKAQLRQREVVDLYNGMCLQGPAGVPGRDGSPGANGIPGTPGIPGRDGFKGEKGECLRESFEESWTPNYKQCSWSSLNYGIDLGKIAECTFTKMRSNSALRVLFSpecifications
collagen triple helix repeat containing 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CTHRC1. | |
CTHRC1 | |
CTHRC1 | |
CTHRC1 (NP_612464, 32 a.a. ∼ 141 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
115908 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_138455 | |
Mouse | |
50 μL | |
Cell Adhesion | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction