Learn More
anti-Ctip2-ChIP Grade, Clone: 25B6, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Supplier: Abcam AB18465100UG

Description
Host Species: Rat
Clone: 25B6
Isotype: IgG
Cross Reactivity: Mouse, Human, Zebrafish
Immunogen: Fusion protein (Human).
Applications:
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
Ctip2-ChIP Grade | |
25B6 | |
Unconjugated | |
Fusion protein (Human). | |
Primary | |
IgG |
Monoclonal | |
0.63-0.86mg/mL | |
Rat | |
100 μg | |
Human, Mouse, Zebrafish |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.