Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CTLA-4, PE, Clone: 63828, R&D Systems™

Catalog No. FAB434P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB434P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB434P Supplier R&D Systems Supplier No. FAB434P
This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen CTLA-4
Applications Flow Cytometry
Classification Monoclonal
Clone 63828
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. XP_001479180
Gene Alias CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Gene Symbols CTLA4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CTLA-4, Ala36-Phe161, Accession # XP_001479180
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1493
Test Specificity Detects mouse CTLA-4 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) CD28, recombinant human CTLA-4, rmICOS, or rmPD-1.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.