Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTLA4 (CD152) Hamster anti-Mouse, Clone: 9H10, MilliporeSigma™

Catalog No. 04963MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-963-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-963-MI Supplier MilliporeSigma Supplier No. 04963
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

Anti-CTLA4 (CD152) Antibody, clone 9H10 is an antibody against CTLA4 (CD152) for use in western blot, FC and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTLA4 (CD152)
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9H10
Conjugate Unconjugated
Formulation Purified hamster monoclonal IgG in buffer containing 0.1MTris-Glycine (pH 7.4) with 150mM NaCl and 0.05% sodium azide.
Gene Accession No. NP_033973.2
Gene Symbols CD, CD152, CD28, CELIAC3, CTLA-4, GSE, ICOS, IDDM12, OTTHUMP00000163781
Host Species Hamster
Immunogen Mouse CTLA-4/human IgG1 fusion protein corresponding to residues surrounding CTLA4.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.