Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

catenin (cadherin-associated protein), beta 1, 88kDa, Mouse, Clone: 1C9, Abnova™

Catalog No. 89000600 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-600 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-600 Supplier Abnova Corporation Supplier No. H00001499M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CTNNB1.

Beta-catenin is an adherens junction protein. Adherens junctions (AJs; also called the zonula adherens) are critical for the establishment and maintenance of epithelial layers, such as those lining organ surfaces. AJs mediate adhesion between cells, communicate a signal that neighboring cells are present, and anchor the actin cytoskeleton. In serving these roles, AJs regulate normal cell growth and behavior. At several stages of embryogenesis, wound healing, and tumor cell metastasis, cells form and leave epithelia. This process, which involves the disruption and reestablishment of epithelial cell-cell contacts, may be regulated by the disassembly and assembly of AJs. AJs may also function in the transmission of the 'contact inhibition' signal, which instructs cells to stop dividing once an epithelial sheet is complete.[supplied by OMIM

Sequence: LFRTEPMAWNETADLGLDIGAQGEPLGYRQDDPSYRSFHSGGYGQDALGMDPMMEHEMGGHHPGADYPVDGLPDLGHAQDLMDGLPPGDSNQLAWFDTDL

Specifications

Antigen catenin (cadherin-associated protein), beta 1, 88kDa
Applications ELISA, Immunofluorescence, In situ PLA, KnockDown, Western Blot
Classification Monoclonal
Clone 1C9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CTNNB1.
Formulation PBS with no preservative; pH 7.4
Gene CTNNB1
Gene Accession No. NM_001904
Gene Alias CTNNB/DKFZp686D02253/FLJ25606/FLJ37923
Gene Symbols CTNNB1
Host Species Mouse
Immunogen CTNNB1 (AAH58926, 682 a.a. ∼ 781 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 1499
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.