Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTNND2, Mouse, Clone: 6E11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CTNND2.
Supplier: Abnova Corporation H00001501M01
Description
Sequence: ISLKERKTDYECTGSNATYHGAKGEHTSRKDAMTAQNTGISTLYRNSYGAPAEDIKHNQVSAQPVPQEPSRKDYETYQPFQNSTRNYDESFFEDQVHHRPPASEYTMHLGSpecifications
CTNND2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001332 | |
CTNND2 | |
CTNND2 (NP_001323, 1081 a.a. ∼ 1190 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
6E11 | |
Mouse monoclonal antibody raised against a partial recombinant CTNND2. | |
CTNND2 | |
GT24/NPRAP | |
Mouse | |
Affinity chromatography | |
RUO | |
1501 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction