Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTPS, Mouse, Clone: 2G7-1D10, Abnova™

Catalog No. 89021228 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-228 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-228 Supplier Abnova Corporation Supplier No. H00001503M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CTPS.

The catalytic conversion of UTP to CTP is accomplished by the enzyme cytidine-5-prime-triphosphate synthetase. The enzyme is important in the biosynthesis of phospholipids and nucleic acids, and plays a key role in cell growth, development, and tumorigenesis. The region to which the CTPS gene has been mapped is the location of breakpoints involved in several tumor types [provided by RefSeq

Sequence: MKYILVTGGVISGIGKGIIASSVGTILKSCGLHVTSIKIDPYINIDAGTFSPYEHGEVFVLDDGGEVDLDLGNYERFLDIRLTKDNNLTTGKIYQYVINKERKGDYLGKTVQVVPHITDAIQEWVMRQALIPVDEDGLEPQVCVIELGGTVGDIESMPFIEAFRQFQFKVKRENFCNIHVSLVPQPSSTGEQKTKPTQNSVRELRGLGLSPDLVVCRCSNPLDTSVKEKISMFCHVEPEQVICVHDVSSIYRVPLLLEEQGVVDYFLRRLDLPIERQPRKMLMKWKEMADRYDRLLETCSIALVGKYTKFSDSYASVIKALEHSALAINHKLEIKYIDSADLEPITSQEEPVRYHEAWQKLCSAHGVLVPGGFGVRGTEGKIQAIAWARNQKKPFLGVCLGMQLAVVEFSRNVLGWQDANSTEFDPTTSHPVVVDMPEHNPGQMGGTMRLGKRRTLFQTKNSVMRKLYGDADYLEERHRHRFEVNPVWKKCLEEQGLKFVGQDVEGERMEIVELEDHPFFVGVQYHPEFLSRPIKPSPPYFGLLLASVGRLSHYLQKGCRLSPRDTYSDRIGSSSPDSEITELKFPSINHD

Specifications

Antigen CTPS
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G7-1D10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CTPS.
Formulation PBS with no preservative; pH 7.4
Gene CTPS
Gene Accession No. BC009408
Gene Symbols CTPS
Host Species Mouse
Immunogen CTPS (AAH09408, 1 a.a. ∼ 591 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1503
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.