Learn More
CTSL2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CTSL2 peptide using ARM Technology.
Supplier: Abnova Corporation H00001515K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may play an important role in corneal physiology. This gene is expressed in colorectal and breast carcinomas but not in normal colon, mammary gland, or peritumoral tissues, suggesting a possible role for this gene in tumor processes. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
CTSL2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
CTSL2 | |
CTSL2 | |
A synthetic peptide of human CTSL2 | |
RUO | |
1515 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
cathepsin L2 | |
CTSL2 | |
CATL2, CTSU, CTSV, MGC125957 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.