Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTSW, Mouse, Clone: 4A8, Abnova™

Catalog No. 89021233 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-233 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-233 Supplier Abnova Corporation Supplier No. H00001521M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CTSW.

The protein encoded by this gene, a member of the peptidase C1 family, is a cysteine proteinase that may have a specific function in the mechanism or regulation of T-cell cytolytic activity. The encoded protein is found associated with the membrane inside the endoplasmic reticulum of natural killer and cytotoxic T-cells. Expression of this gene is up-regulated by interleukin-2. [provided by RefSeq

Sequence: IRGPLRAQDLGPQPLELKEAFKLFQIQFNRSYLSPEEHAHRLDIFAHNLAQAQRLQEEDLGTAEFGVTPFSDLTEEEFGQLYGYRRAAGGVPSMGREIRSEEPEESVPFSCDWRKVAGAISPIKDQKNCNCCWAMAAAGNIETLWRISFWDFVDVSVQELLDCGRCGDGCHGGFVWDAFITVLNNSGLASEKDYPFQGKVRAHRCHPKKYQKVAWIQDFIMLQNNEHRIAQYLATYGPITVTINMKPLQLYRKGVIKATPTTCDPQLVDHSVLLVGFGSVKSEEGIWAETVSSQSQPQPPHPTPYWILKNSWGAQWGEKGYFRLHRGSNTCGITKFPLTARVQKPDMKPRVSCPP

Specifications

Antigen CTSW
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4A8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CTSW.
Formulation PBS with no preservative; pH 7.4
Gene CTSW
Gene Accession No. BC048255
Gene Alias LYPN
Gene Symbols CTSW
Host Species Mouse
Immunogen CTSW (AAH48255, 22 a.a. ∼ 376 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1521
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.