Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Cullin-4A/CUL4A Clone: 2A2.1,

Catalog No. MABC554MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC554MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC554MI Supplier MilliporeSigma™ Supplier No. MABC554
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Cullin-4A/CUL4A Clone: 2A2.1 in Human samples, and it is validated for Western Blotting

Cullin-4A (UniProt Q13619; also known as CUL-4A) is encoded by the CUL4A gene (Gene ID 8451) in human. Polyubiquitination of proteins is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type (really interesting new gene), HECT-type (homologous to E6-AP C-terminus), and U-box type of E3 ligases. Cullin-4 belongs to the cullin family of hydrophobic proteins that confer substrate specificity to the multimeric Cullin-RING E3 ubiquitin ligase (CRL) complexes by acting as scaffold proteins. The human genome encodes 8 cullin proteins (CUL 1, 2, 3, 4A, 4B, 5, 7 and 9). The activity of CRLs is regulated by Cullin Neddylation via the reversible conjugation of a small ubiquitin-type protein known as Neural precursor cell expressed developmentally down-regulated protein 8 (Nedd8). CUL4A interacts with the substrate-binding adaptor DDB1 (DNA damage binding protein 1) via its arc-shaped helical N-terminal domain, and binds the small RING finger protein ROC1 (regulator of cullins 1) via its globular C-terminal domain. Despite the 83% sequence identity and seemingly functional redundancy between CUL4A and CUL4B, much more focus is placed on CUL4A in oncogenesis research.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin-4A/CUL4A
Applications Western Blot
Classification Monoclonal
Clone 2A2.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q13619
Gene Symbols CUL4A
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human Cullin-4A/CUL4A near the C-terminus.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001008895
Test Specificity Clone 2A2.1 is expected to react with both spliced isoforms of human cullin-4A
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.