Learn More
Cutaneous Lymphocyte Antigen Rat anti-Human, Alexa Fluor 647, Clone: HECA-452, BD
Rat Monoclonal Antibody
Supplier:  BD Biosciences 563533
Catalog No. BDB563533
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
The HECA-452 monoclonal antibody specifically reacts with Cutaneous Lymphocyte Associated Antigen (CLA), a carbohydrate domain shared by sialyl Lewis[x] (sLe[x]) and sialyl Lewis[a] (sLe[a]) antigens. It serves as a ligand for selectins including CD62E (E-selectin; ELAM-1). CLA is expressed on high endothelium and on lymphocytes including most T lymphocytes infiltrating cutaneous sites of inflammation. Amongst peripheral blood cells, it is expressed on monocytes and granulocytes and a subset of lymphocytes. The HECA-452 antibody is also reportedly crossreactive with the mouse CLA carbohydrate epitope that is transiently expressed by PSGL-1/CD162 on activated T cells. A number of studies suggest that CLA plays an important role in supporting leucocyte adhesive interactions and migration into extravascular tissues during inflammation.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.