Learn More
Description
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq
Specifications
Specifications
| Antigen | cut-like homeobox 1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2D10 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant CUTL1. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CUX1 |
| Gene Accession No. | NM_001913 |
| Gene Alias | CASP/CDP/CDP/Cut/CDP1/COY1/CUTL1/CUX/Clox/Cux/CDP/GOLIM6/Nbla10317/p100/p110/p200/p75 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.