Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CWC15 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524738

Catalog No. PIPA524738

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

CWC15 involved in pre-mRNA splicing.
Specifications

Specifications

CWC15
Polyclonal
PBS with 0.09% sodium azide
Q9P013
C11orf5, Cwf15, HSPC148, ORF5, CWC15 homolog, protein CWC15 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100775065, 51503
Immunohistochemistry, Western Blot
Unconjugated
CWC15
Liquid
CWC15
KLH conjugated synthetic peptide between 123-152 amino acids from the central region of human CWC15
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit