Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CX3CL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523062
Description
Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-a in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.
Specifications
CX3CL1 | |
Polyclonal | |
Unconjugated | |
Cx3cl1 | |
A-152E5.2; AB030188; ABCD-3; Acc1; AI848747; C3Xkine; chemokine (C-X3-C motif) ligand 1; CX3C; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1; C-X3-C motif chemokine ligand 1; Cx3cl1; CXC3; CXC3C; D8Bwg0439e; Fkn; Fractalkine; neurotactin; NTN; NTT; Processed fractalkine; SCYD1; small inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, neurotactin); small inducible cytokine subfamily D (Cys-X3-Cys), member-1; small inducible cytokine subfamily D, 1; small-inducible cytokine D1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20312, 6376 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1.15 mg/mL | |
PBS with 30% glycerol and 0.05% sodium azide | |
O35188, P78423 | |
Cx3cl1 | |
A synthetic peptide made to an internal region of the human CX3CL protein (within residues 20-150). | |
100 μL | |
Primary | |
Gerbil, Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction