Learn More
anti-CX3CR1, PerCP, Clone: 528728, R&D Systems™
Description
Host Species: Mouse
Clone: 528728
Isotype: IgG1
Species Reactivity: Hu
Immunogen: E. coli-derived recombinant human CX3CR1. Met1-Thr31, Leu91-Lys103, Thr168-Thr195, Lys257-Leu273
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | CX3CR1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 528728 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Gene Accession No. | NP_001328 |
| Gene Alias | Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1 |
| Gene Symbols | CX3CR1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.