Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CX3CR1, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127533
Description
Receptor for the CX3C chemokine fractalkine and mediates both its adhesive and migratory functions. Acts as coreceptor with CD4 for HIV-1 virus envelope protein (in vitro). Isoform 2 and isoform 3 seem to be more potent HIV coreceptors than isoform 1.
PA1-27533 detects CX3CR1 from human samples.PA1-27533 may be successfully used in ELISA and Western blot applications.The PA1-27493 immunogen is a synthetic peptide corresponding to human CX3C chemokine receptor 1.
Specifications
CX3CR1 | |
Polyclonal | |
CX3CR1 | |
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
1524 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
ELISA, Western Blot | |
Unconjugated | |
P49238 | |
CX3CR1 | |
Synthetic peptide corresponding to human CX3C chemokine receptor 1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction