Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL11, Mouse, Clone: 3C1, Abnova™

Catalog No. 89123115 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-115 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-115 Supplier Abnova Corporation Supplier No. H00006373M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CXCL11.

Chemokines are a group of small (approximately 8 to 14kDa), mostly basic, structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane, G protein-coupled receptors. Chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies, CXC and CC. This gene is a CXC member of the chemokine superfamily. Its encoded protein induces a chemotactic response in activated T-cells and is the dominant ligand for CXC receptor-3. The gene encoding this protein contains 4 exons and at least three polyadenylation signals which might reflect cell-specific regulation of expression. IFN-gamma is a potent inducer of transcription of this gene. [provided by RefSeq

Sequence: MSVKGMAIALAVILCATVVQGFPMFKRGRCLCIGPGVKAVKVADIEKASIMYPSNNCDKIEVIITLKENKGQRCLNPKSKQARLIIKKVERKNF

Specifications

Antigen CXCL11
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 3C1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant CXCL11.
Formulation ascites with no preservative
Gene CXCL11
Gene Accession No. BC005292
Gene Alias H174/I-TAC/IP-9/IP9/MGC102770/SCYB11/SCYB9B/b-R1
Gene Symbols CXCL11
Host Species Mouse
Immunogen CXCL11 (AAH05292, 1 a.a. ∼ 94 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6373
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.