Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL12, Mouse, Clone: 2E2, Abnova™

Catalog No. 89123286 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-286 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-286 Supplier Abnova Corporation Supplier No. H00006387M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CXCL12.

For background information on chemokines, see CXCL1 (MIM 155730). Stromal cell-derived factors 1-alpha and 1-beta are small cytokines that belong to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF (see MIM 191160), or IL1 (see MIM 147760). The intercrines are characterized by the presence of 4 conserved cysteines which form 2 disulfide bonds. They can be classified into 2 subfamilies. In the CC subfamily, which includes beta chemokine, the cysteine residues are adjacent to each other. In the CXC subfamily, which includes alpha chemokine, they are separated by an intervening amino acid. The SDF1 proteins belong to the latter group.[supplied by OMIM

Sequence: MNAKVVVVLVLVLTALCLSDGKPVSLSYRCPCRFFESHVARANVKHLKILNTPNCALQIVARLKNNNRQVCIDPKLKWIQEYLEKALNK

Specifications

Antigen CXCL12
Applications ELISA
Classification Monoclonal
Clone 2E2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CXCL12.
Formulation PBS with no preservative; pH 7.4
Gene CXCL12
Gene Accession No. BC039893
Gene Alias PBSF/SCYB12/SDF-1a/SDF-1b/SDF1/SDF1A/SDF1B/TLSF-a/TLSF-b/TPAR1
Gene Symbols CXCL12
Host Species Mouse
Immunogen CXCL12 (AAH39893.1, 1 a.a. ∼ 89 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6387
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.