Learn More
Description
For background information on chemokines, see CXCL1 (MIM 155730). Stromal cell-derived factors 1-alpha and 1-beta are small cytokines that belong to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF (see MIM 191160), or IL1 (see MIM 147760). The intercrines are characterized by the presence of 4 conserved cysteines which form 2 disulfide bonds. They can be classified into 2 subfamilies. In the CC subfamily, which includes beta chemokine, the cysteine residues are adjacent to each other. In the CXC subfamily, which includes alpha chemokine, they are separated by an intervening amino acid. The SDF1 proteins belong to the latter group.[supplied by OMIM
Specifications
Specifications
| Antigen | CXCL12 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4D8 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant CXCL12. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CXCL12 |
| Gene Accession No. | BC039893 |
| Gene Alias | PBSF/SCYB12/SDF-1a/SDF-1b/SDF1/SDF1A/SDF1B/TLSF-a/TLSF-b/TPAR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.