Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCL6/GCP-2, APC, Clone: 60910, R&D Systems™

Catalog No. IC333A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC333A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC333A Supplier R&D Systems Supplier No. IC333A
This item is not returnable. View return policy

Applications:

Flow Cytometry (Intracellular Staining)

Specifications

Antigen CXCL6/GCP-2
Applications Flow Cytometry
Classification Monoclonal
Clone 60910
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P80162
Gene Alias chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2), Chemokine alpha 3, CKA-3Granulocyte chemotactic protein 2, GCP-2, GCP2member 6 (granulocytechemotactic protein 2), GCP-2Small-inducible cytokine B6, Small inducible cytokine subfamily B (Cys-X-Cys), member b
Gene Symbols CXCL6
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL6/GCP-2, Val40-Asn114, Accession # P80162
Purification Method Protein A or G purified from ascites
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6372
Test Specificity Detects human CXCL6/GCP-2 in ELISAs and Western blots. In sandwich immunoassays, less than 0.04% cross-reactivity with recombinant human (rh) NAP-2, rhGRO alpha , rhGRO beta , rhBLC, rhENA-78, recombinant mouse (rm) LIX, rmMIP-2, rhIL-8, rhIP-10, rhMIG, rhSDF-1 alpha , and rhSDF-1 beta is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.