Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL8/IL-8 Mouse anti-Porcine, Clone: 105115, R&D Systems™

Catalog No. MAB535
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB535 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB535 Supplier R&D Systems Supplier No. MAB535
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL8/IL-8 Monoclonal antibody MAB535 Specifically detects CXCL8/IL-8 in Porcine samples. It is validated for Neutralization, Western Blotting

Specifications

Antigen CXCL8/IL-8
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 105115
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.2-1.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. CAA43461
Gene Alias 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-
Gene Symbols IL8
Host Species Mouse
Immunogen E. coli-derived recombinant porcine IL-8/CXCL8 Ala26-Gln104 Accession # CAA43461
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3576
Test Specificity Detects porcine IL-8/CXCL8 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) CXCL8 or rhCXCL1, 2, 3, 5, 6, 7, 9, 10, 11, 12, 13, recombinant mouse CXCL1, 2, 6, 9, 10, 12, 13, recombinant rat CXCL1, 2 or 3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Pig
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.