Learn More
anti-CXCR1/IL-8 RA, APC, Clone: 42705, R&D Systems™
Description
Host Species: Mouse
Clone: 42705
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA. Met1-Leu350
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | CXCR1/IL-8 RA |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 42705 |
| Conjugate | APC |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Gene Accession No. | AAA59159 |
| Gene Alias | CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RA, IL-8RA, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A |
| Gene Symbols | CXCR1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.