Learn More
anti-CXCR2/IL-8 RB, PerCP, Clone: 242216, R&D Systems™
Description
Host Species: Rat
Clone: 242216
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: C6 rat glioma cell line transfected with mouse CXCR2. Met1-Leu359
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | CXCR2/IL-8 RB |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 242216 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Gene Accession No. | P35343 |
| Gene Alias | beta, CD182, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, IL8RB, IL-8RB, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B |
| Gene Symbols | CXCR2 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.