Learn More
anti-CXCR3, Alexa Fluor 700, Clone: 220803, R&D Systems™
Description
Host Species: Rat
Clone: 220803
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: CHO Chinese hamster ovary cell line transfected with mouse CCR3. Met1-Leu367
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | CXCR3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 220803 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Gene Accession No. | AAC40163 |
| Gene Alias | CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR |
| Gene Symbols | CXCR3 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.