Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CXCR3 APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 6 publications
$138.04 - $474.44
Specifications
Antigen | CXCR3 |
---|---|
Clone | 220803 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1685A025
|
R&D Systems
FAB1685A025 |
25 Tests |
Each for $138.04
|
|
|||||
FAB1685A100
|
R&D Systems
FAB1685A100 |
100 Tests |
Each for $474.44
|
|
|||||
Description
CXCR3 Monoclonal specifically detects CXCR3 in Mouse samples. It is validated for Flow Cytometry.Specifications
CXCR3 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
Mouse | |
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
CXCR3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse CXCR3. |
220803 | |
Flow Cytometry | |
APC | |
Rat | |
AAC40163 | |
2833 | |
CHO Chinese hamster ovary cell line transfected with mouse CXCR3 Met1-Leu367 Accession # AAC40163 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title