Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR4, APC, Clone: 247506, R&D Systems™

Catalog No. FAB21651A10
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB21651A10 100 Tests
FAB21651A02 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB21651A10 Supplier R&D Systems Supplier No. FAB21651A100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Host Species: Rat
Clone: 247506
Isotype: IgG2B
Species Reactivity: Ms
Immunogen: Y3 rat myeloid cell line transfected with mouse CXCR4. Met1-Ser359

Applications:

Flow Cytometry

Specifications

Antigen CXCR4
Applications Flow Cytometry
Classification Monoclonal
Clone 247506
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P70658
Gene Alias CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor
Gene Symbols CXCR4
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with mouse CXCR4 Met1-Ser359 Accession # P70658
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 7852
Test Specificity Detects mouse CXCR4 transfectants. Does not stain irrelevant transfectants.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.