Learn More
CXCR5 Rat anti-Human, Alexa Fluor 488, Clone: RF8B2, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 558112
Description
The monoclonal antibody RF8B2 reacts with the human CXC chemokine receptor, CXCR5. CXCR5 (aka BLR-1 NLR and MDR15), a seven transmembrane, G-protein-coupled receptor, is the specific receptor for CXC chemokine, CXCL13/BLC/BCA-1. In peripheral blood, CXCR5 expression is restricted to B lymphocytes and a small subset of CD4+ and CD8+ lymphocytes. The restricted expression pattern of CXCR5 on B cells suggested that this receptor might function as a regulator of B cell migration. Stimulation of T cells with anti-CD3 monoclonal antibody leads to the down-regulation of CXCR5.
Host Species: Rat
Clone: RF8B2
Isotype: IgG2b, κ
Cross Reactivity: Human
Flow Cytometry, Immunofluorescence

Specifications
CXCR5 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Human CXCR5 | |
0.1 mg | |
Immunology | |
Human | |
IgG2b κ |
Flow Cytometry | |
RF8B2 | |
Alexa Fluor 488 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.