Learn More
CXCR5 Rat anti-Mouse, APC, Clone: 2G8, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 560615
Description
The monoclonal antibody 2G8 reacts with the mouse CXC chemokine receptor, CXCR5. CXCR5 (a.k.a. BLR1, NLR and MDR15), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CXC chemokines, CXCL13/BLC/BCA-1. The expression of CXCR5 has been detected in spleen, lymph nodes, tonsils, brain, bone marrow, T cells, B cells, cerebrum, cerebellum, hippcampus and pituitary. In mouse spleen, CXCR5 was strictly expressed by mature B cells and a small subset of T lymphocytes. The immunogen used to
generate 2G8 hybridoma was a recombinant protein containing N-terminal amino acids of mouse CXCR5 (GST-NmBLR1).
Flow Cytometry

Specifications
CXCR5 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse CXCR5 | |
50 μg | |
Immunology | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
2G8 | |
APC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.