Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCR7/RDC-1 Fluorescein-conjugated Antibody, R&D Systems™

Catalog No. FAB42271F10
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB42271F10 100 Tests
FAB42271F02 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB42271F10 Supplier R&D Systems Supplier No. FAB42271F100
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR7/RDC-1
Applications Flow Cytometry
Classification Monoclonal
Clone 358426
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAA62370
Gene Alias ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Gene Symbols CXCR7
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR7/RDC-1 Met1-Lys362 (Gly131Ser) Accession # AAA62370
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 57007
Test Specificity Detects human CXCR7/RDC-1 in direct ELISAs. In flow cytometry, reacts specifically with five distinct human CXCR7 transfectants, but does not react with their respective parental lines or mouse CXCR7 transfectants. In flow cytometry, also reacts with monocytes expressing CXCR7, but does not react with MCF-7 cells which have been reported to have surface-expressing CXCR7 using clone 11G8. Due to the conflicting reports published, use of clone 358426 may result in an underestimation of CXCR7 expression on certain cell types.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.