Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB5R3, Mouse, Clone: 2A9, Abnova™

Catalog No. 89021298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-298 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-298 Supplier Abnova Corporation Supplier No. H00001727M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CYB5R3.

This gene encodes cytochrome b5 reductase, which includes a membrane-bound form in somatic cells (anchored in the endoplasmic reticulum, mitochondrial and other membranes) and a soluble form in erythrocytes. The membrane-bound form exists mainly on the cytoplasmic side of the endoplasmic reticulum and functions in desaturation and elongation of fatty acids, in cholesterol biosynthesis, and in drug metabolism. The erythrocyte form is located in a soluble fraction of circulating erythrocytes and is involved in methemoglobin reduction. The membrane-bound form has both membrane-binding and catalytic domains, while the soluble form has only the catalytic domain. These two forms are resulted from alternative splicing of the gene. Mutations in this gene cause methemoglobinemias. [provided by RefSeq

Sequence: FAIRPDKKSNPIIRTVKSVGMIAGGTGITPMLQVIRAIMKDPDDHTVCHLLFANQTEKDILLRPELEELRNKHSARFKLWYTLDRAPEAWDYGQGF

Specifications

Antigen CYB5R3
Applications ELISA
Classification Monoclonal
Clone 2A9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CYB5R3.
Formulation PBS with no preservative; pH 7.4
Gene CYB5R3
Gene Accession No. BC004821
Gene Alias B5R/DIA1
Gene Symbols CYB5R3
Host Species Mouse
Immunogen CYB5R3 (AAH04821.1, 157 a.a. ∼ 252 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1727
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.