Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ anti-cyclic AMP, Polyclonal

Catalog No. AAJ64001AMO
Change view
Click to view available options
Quantity:
1000 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64001AMO 1000 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64001AMO Supplier Thermo Scientific™ Supplier No. J64001AMO

Rabbit Polyclonal Antibody

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Antigen cyclic AMP
Applications Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Quantity 1000 Tests
Primary or Secondary Primary
Content And Storage 2-8°C
Form Lyophilized

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.