Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A Mouse anti-Human, Unlabeled, Clone: BF683, BD

Catalog No. BDB554174 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554174 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554174 Supplier BD Biosciences Supplier No. 554174
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins (regulatory subunits) bind to cdks (catalytic subunits) to form complexes that regulate the progression of the cell cycle. The main cyclin-cdks complexes formed in vertebrate cells are cyclin D-cdk4 (G0/G1), cyclin E-cdk2 (G1/S), cyclin A-cdk2 (S) and cyclin B1-cdk1 (G2/M). These complexes are regulated by activating and inhibitory phosphorylation events, as well as by interactions with small regulatory proteins including p21 and p27Kip1. Specific substrates for cdk-cyclin complexes include nuclear lamins, histones, oncogenes (e.g., c-abl and SV40 large T-Ag), tumor suppressor genes (e.g., retinoblastoma protein, Rb), nucleolin and others. Cyclin A is involved in both S-phase and G2/M transitions of the cell cycle through its association with cdk2 and cdk1, respectively. Cyclin A may also form a complex with the adenovirus oncoprotein E1A which has DNA binding activity. Human cyclin A has been reported to migrate between 54-60kDa by SDS-PAGE and clone BF683 reportedly does not cross-react with mouse, rat or mink cyclin A.

Host Species: Mouse
Clone: BF683
Isotype: IgE
Species Reactivity: Human
Immunogen: Human Cyclin A Recombinant Protein
Formula Weight [Chemical]: 54-60kDa

Flow Cytometry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A
Applications Western Blot
Classification Monoclonal
Clone BF683
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Cyclin A Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgE
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.