Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B1 Mouse anti-Human, Mouse, Phycoerythrin, Clone: GNS3 (8A5D12), MilliporeSigma™

Catalog No. FCMAB102PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB102PMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB102PMI Supplier MilliporeSigma Supplier No. FCMAB102P

Mouse Monoclonal Antibody

This Milli-Mark™ Anti-Cyclin B1-PE Antibody, clone GNS3 (8A5D12) is validated for use in western blot, immunoprecipitation, FC for the detection of Cyclin B1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin B1-PE
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GNS3 (8A5D12)
Conjugate PE
Formulation Protein G purified mouse IgGs conjugated to phycoerythrin in storage buffer PBS, 15mg/mL BSA and 0.1% sodium azide with out Glycerol.
Gene Accession No. NM_031966
Gene Symbols G2/mitotic-specific cyclin B1, cyclin B1
Host Species Mouse
Immunogen Recombinant human cyclin B1
Purification Method Protein G purfied
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.