Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B1 Mouse, FITC, Clone: GNS-1, BD

Catalog No. BDB554108 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554108 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554108 Supplier BD Biosciences Supplier No. 554108
Add to Cart
Add to Cart

Includes

FITC Mouse Anti-Human Cyclin B1 and FITC Mouse IgG1, kappa Isotype Control

Mouse Monoclonal Antibody

Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins (regulatory subunits) bind to cdks (catalytic subunits) to form complexes that regulate the progression of the cell cycle. The main cyclin-cdks complexes formed in vertebrae cells are cyclin-cdks complexes that regulate the progression of the cell cycle. The main cyclin-cdks complexes formed in vertebrae cells are cyclin D-cdk4 (G0/G1), cyclin E-cdk2 (G1/S), cyclin A-cdk2 (S) and cyclin B1-cdk1 (G2/M). These complexes are regulated by activating and inhibitory phosphorylation events, as well as by interactions with small regulatory proteins including p21 and p27[Kip1]. Specific substrates for cdk-cyclin complexes include nuclear lamins, histones, oncogenes (e.g., c-abl and SV40 large T-Ag), tumor suppressor genes (e.g., retinoblastoma protein, Rb) nucleolin and others. Cyclin B1 is a mitotic cyclin complex; expression is normally low in G0/G1, increases in S, and is maximal during G2/M. Cyclin B1 is rapidly degraded at the end of mitosis and is required for cells to exit from mitosis.
GNS-1 recognizes an epitope between amino acids 1-21 of human Cyclin B1. It crossreacts with hamster and mouse cyclin B1. Recombinant human Cyclin B1 was used as immunogen. The antibody was originally characterized by western blot analysis, immunoprecipitation, immunohistochemical staining of acetone-fixed, frozen tissue sections and of formalin-fixed, paraffin-embedded tissue sections, indirect immunofluorescence microscopy of cultured cells and flow cytometry. MOPC-1 is used as a mouse IgG1 isotype (negative) control. The MOPC-21 antibody has unknown specificity. The GNS-1 and MOPC-21 FITC conjugates are matched in F/P ratios. The optimum F/P ratio was experimentally determined by flow cytometric analysis. MOLT4 human leukemia cells (ATCC CRL-1582) are recommended as a positive control for this application.

Host Species: Mouse
Clone: GNS-1, MOPC-21
Isotype: IgG1, IgG1 κ
Species Reactivity: Human
Immunogen: Human Cyclin B1 Recombinant Protein

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin B1
Applications Flow Cytometry
Classification Monoclonal
Clone GNS-1
Conjugate FITC
Host Species Mouse
Immunogen Human Cyclin B1 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Includes FITC Mouse Anti-Human Cyclin B1 (100 tests), FITC Mouse IgG1, kappa Isotype Control (100 tests)
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.