Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D1 Mouse anti-Human, Unlabeled, Clone: DCS-6, BD

Catalog No. BDB556470 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556470 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556470 Supplier BD Biosciences Supplier No. 556470
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclins and cyclin-dependent kinases (Cdks) have been shown to be subunits of cell cycle dependent protein kinases that regulate key events during the progression of the cell cycle and are evolutionarily highly conserved. Specific substrates for Cdk/cyclin kinases include nuclear lamins, histones, oncogenes (c-src, c-abl, SV40 large-T), tumor suppressor genes (the retinoblastoma protein [Rb] and p53), nucleolin, RNA polymerase II and others. D-type cyclins are involved in regulating the passage of mammalian cells through G1. In SDS-PAGE, D-type cyclins migrate at the following molecular weights: cyclin D1 (36kDa), cyclin D2 (35kDa), and cyclin D3 [31 and 34kDa (doublet)]. Rodent cyclin D1 homologues (Cyl1) have been reported to typically migrate as a 36kDa doublet. The mouse anti-human cyclin D1 antibody (clone DCS-6) recognizes human cyclin D1 (36kDa) and has been reported to crossreact with rat and mouse cyclin D1 homologues (Cyl1). It does not crossreact with human cyclins D2 and D3.

Host Species: Mouse
Clone: DCS-6
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Human Full-length Cyclin D1 Recombinant Protein

Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin D1
Applications Western Blot
Classification Monoclonal
Clone DCS-6
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Full-length Cyclin D1 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.