Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D1 Mouse anti-Human, Unlabeled, Clone: G124-326, BD

Catalog No. BDB554180 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554180 0.1 mg
BDB554181 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB554180 Supplier BD Biosciences Supplier No. 554180
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins contain a conserved amino acid sequence motif, the cyclin box, which allows their binding to cdks to form active complexes that regulate the progression of the cell cycle. The synthesis and degradation of cyclins is tightly controlled in a cell cycle specific manner. Several classes of cyclins (A-E) have been described. Cyclins have been placed into functional groups as follows: Group 1 (cyclins A, B, D1, D2, D3, E and F) functions primarily in cell cycle reguation; Group 2 (cyclins C and H) also plays a role in transcriptional regulation; Group 3 (cyclins G1, G2 and I) may play a role distinct from other cyclins. Specific substrates for cyclin-cdk complexes include nuclear lamins, histones, oncogenes (c-src, c-abl, SV40 large-T Ag), tumor suppressor genes (Rb and p53), and others. The D-type cyclins are involved in regulating the passage of mammalian cells through G1. The reduced molecular weights of cyclin D1 is ∽36kda. G124-326 recognizes human cyclin D1 and cross-reacts with the mouse homolog (Cyl1) of human cyclin D1. It does not cross-react with human cyclins D2 and D3. Recombinant full-length human cyclin D1 was used as immunogen.

Host Species: Mouse
Clone: G124-326
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant full-length human cyclin D1
Formula Weight [Chemical]: 36kDa

Flow Cytometry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin D1
Applications Western Blot
Classification Monoclonal
Clone G124-326
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant full-length human cyclin D1
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.