Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB610279 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610279 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610279 Supplier BD Biosciences Supplier No. 610279
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Several classes of cyclins (A-E) have been described. These proteins act as regulatory subunits for cyclin-dependent kinases (cdks). The synthesis and degradation of cyclins is tightly controlled in a cell cycle specific manner. There are at least 3 different D-type cyclins whose relative levels vary among cell types. Cyclin D3 is known to associate with Cdk5 and weakly with Cdk2. Cdk4 is preferentially bound by Cyclin D1. However, in T cells which do not express Cyclin D1, Cdk4 associates with Cyclins D2 and D3. While bound to cdks, the D-type cyclins also associate with the polymerase-δ subunit, PCNA. Cyclin D3 has also been shown to bind pRb in vitro. This suggests that pRb may be an in vivo substrate of D-type cyclins.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin D3
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.