Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin E Mouse anti-Human, Clone: HE12, MilliporeSigma™

Catalog No. 05363MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-363-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-363-MI Supplier MilliporeSigma Supplier No. 05363
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cyclin E Antibody, clone HE12 is a high quality Mouse Monoclonal Antibody for the detection of Cyclin E and has been validated in immunoprecipitation, western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin E
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HE12
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing 0.05% sodium azide
Gene Accession No. NM_001238, NM_057182
Gene Symbols CCNE1, CCNE
Host Species Mouse
Immunogen Recombinant human cyclin E
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2 years at −20°C or 6 months at 4°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.