Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin F Mouse anti-Human, Unlabeled, Clone: B74-2, BD

Catalog No. BDB556598 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556598 Supplier BD Biosciences Supplier No. 556598

Mouse Monoclonal Antibody

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin F
Applications Western Blot
Classification Monoclonal
Clone B74-2
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.