Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin I2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535081
Description
CCNI2 belongs to the cyclin family.
Specifications
| Cyclin I2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q6ZMN8 | |
| CCNI2 | |
| KLH conjugated synthetic peptide between 92-121 amino acids from the central region of human CCNI2 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| CCNI2 | |
| CCNI2; Cyclin-I2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 645121 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction